Assay Details
| Gene Symbol: LOC100762363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649659.1 | XP_007647849.1 | 10 - 11 | 1361 | 87 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100762363| Gene Symbol: LOC100762363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649659.1 | XP_007647849.1 | 10 - 11 | 1361 | 87 | |