Assay Details
| Gene Symbol: Pdia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641914.1 | XP_007640104.1 | 10 - 11 | 1345 | 58 | |
| GenBank mRNA | JI864823.1 | - | 10 - 11 | 1345 | 58 | |
More Information
Set Membership: |
|

Pdia6
Genomic Map