Assay Details
| Gene Symbol: Eci2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007612706.1 | XP_007610896.1 | 7 - 8 | 899 | 62 | |
| XM_007612716.1 | XP_007610906.1 | 7 - 8 | 825 | 62 | ||
| GenBank mRNA | JI863995.1 | - | 6 - 7 | 742 | 62 | |
More Information
Set Membership: |
|

Eci2
Genomic Map