Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 66 - 67 | 17377 | 94 | |
| XM_007645556.1 | XP_007643746.1 | 66 - 67 | 17377 | 94 | ||
| XM_007645557.1 | XP_007643747.1 | 65 - 66 | 17314 | 94 | ||
| XM_007645558.1 | XP_007643748.1 | 61 - 62 | 11281 | 94 | ||
| XM_007645559.1 | XP_007643749.1 | 61 - 62 | 11281 | 94 | ||
| XM_007645560.1 | XP_007643750.1 | 60 - 61 | 11218 | 94 | ||
| XM_007645561.1 | XP_007643751.1 | 10 - 11 | 6005 | 94 | ||
| GenBank mRNA | JI895671.1 | - | 5 - 6 | 762 | 94 | |
| JP043055.1 | - | 5 - 6 | 762 | 94 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map