Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 71 - 72 | 18149 | 96 | |
| XM_007645556.1 | XP_007643746.1 | 71 - 72 | 18149 | 96 | ||
| XM_007645557.1 | XP_007643747.1 | 70 - 71 | 18086 | 96 | ||
| XM_007645558.1 | XP_007643748.1 | 66 - 67 | 12053 | 96 | ||
| XM_007645559.1 | XP_007643749.1 | 66 - 67 | 12053 | 96 | ||
| XM_007645560.1 | XP_007643750.1 | 65 - 66 | 11990 | 96 | ||
| XM_007645561.1 | XP_007643751.1 | 15 - 16 | 6777 | 96 | ||
| GenBank mRNA | JI895671.1 | - | 10 - 11 | 1534 | 96 | |
| JP043055.1 | - | 10 - 11 | 1534 | 96 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map