Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 74 - 75 | 18601 | 78 | |
| XM_007645556.1 | XP_007643746.1 | 74 - 75 | 18601 | 78 | ||
| XM_007645557.1 | XP_007643747.1 | 73 - 74 | 18538 | 78 | ||
| XM_007645558.1 | XP_007643748.1 | 69 - 70 | 12505 | 78 | ||
| XM_007645559.1 | XP_007643749.1 | 69 - 70 | 12505 | 78 | ||
| XM_007645560.1 | XP_007643750.1 | 68 - 69 | 12442 | 78 | ||
| XM_007645561.1 | XP_007643751.1 | 18 - 19 | 7229 | 78 | ||
| GenBank mRNA | JI895671.1 | - | 13 - 14 | 1986 | 78 | |
| JP043055.1 | - | 13 - 14 | 1986 | 78 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map