Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 75 - 76 | 18703 | 85 | |
| XM_007645556.1 | XP_007643746.1 | 75 - 76 | 18703 | 85 | ||
| XM_007645557.1 | XP_007643747.1 | 74 - 75 | 18640 | 85 | ||
| XM_007645558.1 | XP_007643748.1 | 70 - 71 | 12607 | 85 | ||
| XM_007645559.1 | XP_007643749.1 | 70 - 71 | 12607 | 85 | ||
| XM_007645560.1 | XP_007643750.1 | 69 - 70 | 12544 | 85 | ||
| XM_007645561.1 | XP_007643751.1 | 19 - 20 | 7331 | 85 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map