Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 77 - 78 | 19026 | 80 | |
| XM_007645556.1 | XP_007643746.1 | 77 - 78 | 19026 | 80 | ||
| XM_007645557.1 | XP_007643747.1 | 76 - 77 | 18963 | 80 | ||
| XM_007645558.1 | XP_007643748.1 | 72 - 73 | 12930 | 80 | ||
| XM_007645559.1 | XP_007643749.1 | 72 - 73 | 12930 | 80 | ||
| XM_007645560.1 | XP_007643750.1 | 71 - 72 | 12867 | 80 | ||
| XM_007645561.1 | XP_007643751.1 | 21 - 22 | 7654 | 80 | ||
| GenBank mRNA | JI892635.1 | - | 1 - 2 | 34 | 80 | |
| JP062623.1 | - | 1 - 2 | 34 | 80 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map