Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 78 - 79 | 19238 | 89 | |
| XM_007645556.1 | XP_007643746.1 | 78 - 79 | 19238 | 89 | ||
| XM_007645557.1 | XP_007643747.1 | 77 - 78 | 19175 | 89 | ||
| XM_007645558.1 | XP_007643748.1 | 73 - 74 | 13142 | 89 | ||
| XM_007645559.1 | XP_007643749.1 | 73 - 74 | 13142 | 89 | ||
| XM_007645560.1 | XP_007643750.1 | 72 - 73 | 13079 | 89 | ||
| XM_007645561.1 | XP_007643751.1 | 22 - 23 | 7866 | 89 | ||
| GenBank mRNA | JI892635.1 | - | 2 - 3 | 246 | 89 | |
| JP062623.1 | - | 2 - 3 | 246 | 89 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map