Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 79 - 80 | 19362 | 62 | |
| XM_007645556.1 | XP_007643746.1 | 79 - 80 | 19362 | 62 | ||
| XM_007645557.1 | XP_007643747.1 | 78 - 79 | 19299 | 62 | ||
| XM_007645558.1 | XP_007643748.1 | 74 - 75 | 13266 | 62 | ||
| XM_007645559.1 | XP_007643749.1 | 74 - 75 | 13266 | 62 | ||
| XM_007645560.1 | XP_007643750.1 | 73 - 74 | 13203 | 62 | ||
| XM_007645561.1 | XP_007643751.1 | 23 - 24 | 7990 | 62 | ||
| GenBank mRNA | JI892635.1 | - | 3 - 4 | 370 | 62 | |
| JP062623.1 | - | 3 - 4 | 370 | 62 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map