Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 80 - 81 | 19539 | 112 | |
| XM_007645556.1 | XP_007643746.1 | 80 - 81 | 19539 | 112 | ||
| XM_007645557.1 | XP_007643747.1 | 79 - 80 | 19476 | 112 | ||
| XM_007645558.1 | XP_007643748.1 | 75 - 76 | 13443 | 112 | ||
| XM_007645559.1 | XP_007643749.1 | 75 - 76 | 13443 | 112 | ||
| XM_007645560.1 | XP_007643750.1 | 74 - 75 | 13380 | 112 | ||
| XM_007645561.1 | XP_007643751.1 | 24 - 25 | 8167 | 112 | ||
| GenBank mRNA | JI892635.1 | - | 4 - 5 | 548 | 112 | |
| JP062623.1 | - | 4 - 5 | 547 | 112 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map