Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 86 - 87 | 20546 | 69 | |
| XM_007645556.1 | XP_007643746.1 | 86 - 87 | 20546 | 69 | ||
| XM_007645557.1 | XP_007643747.1 | 85 - 86 | 20483 | 69 | ||
| XM_007645558.1 | XP_007643748.1 | 81 - 82 | 14450 | 69 | ||
| XM_007645559.1 | XP_007643749.1 | 81 - 82 | 14450 | 69 | ||
| XM_007645560.1 | XP_007643750.1 | 80 - 81 | 14387 | 69 | ||
| XM_007645561.1 | XP_007643751.1 | 30 - 31 | 9174 | 69 | ||
| GenBank mRNA | JI890392.1 | - | 4 - 5 | 561 | 69 | |
| JP042596.1 | - | 4 - 5 | 561 | 69 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map