Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 87 - 88 | 20674 | 83 | |
| XM_007645556.1 | XP_007643746.1 | 87 - 88 | 20674 | 83 | ||
| XM_007645557.1 | XP_007643747.1 | 86 - 87 | 20611 | 83 | ||
| XM_007645558.1 | XP_007643748.1 | 82 - 83 | 14578 | 83 | ||
| XM_007645559.1 | XP_007643749.1 | 82 - 83 | 14578 | 83 | ||
| XM_007645560.1 | XP_007643750.1 | 81 - 82 | 14515 | 83 | ||
| XM_007645561.1 | XP_007643751.1 | 31 - 32 | 9302 | 83 | ||
| GenBank mRNA | JI890392.1 | - | 5 - 6 | 689 | 83 | |
| JP042596.1 | - | 5 - 6 | 689 | 83 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map