Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 91 - 92 | 21283 | 77 | |
| XM_007645556.1 | XP_007643746.1 | 91 - 92 | 21283 | 77 | ||
| XM_007645557.1 | XP_007643747.1 | 90 - 91 | 21220 | 77 | ||
| XM_007645558.1 | XP_007643748.1 | 86 - 87 | 15187 | 77 | ||
| XM_007645559.1 | XP_007643749.1 | 86 - 87 | 15187 | 77 | ||
| XM_007645560.1 | XP_007643750.1 | 85 - 86 | 15124 | 77 | ||
| XM_007645561.1 | XP_007643751.1 | 35 - 36 | 9911 | 77 | ||
| GenBank mRNA | JI890392.1 | - | 9 - 10 | 1298 | 77 | |
| JP042596.1 | - | 9 - 10 | 1298 | 77 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map