Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 92 - 93 | 21451 | 84 | |
| XM_007645556.1 | XP_007643746.1 | 92 - 93 | 21451 | 84 | ||
| XM_007645557.1 | XP_007643747.1 | 91 - 92 | 21388 | 84 | ||
| XM_007645558.1 | XP_007643748.1 | 87 - 88 | 15355 | 84 | ||
| XM_007645559.1 | XP_007643749.1 | 87 - 88 | 15355 | 84 | ||
| XM_007645560.1 | XP_007643750.1 | 86 - 87 | 15292 | 84 | ||
| XM_007645561.1 | XP_007643751.1 | 36 - 37 | 10079 | 84 | ||
| GenBank mRNA | JI890392.1 | - | 10 - 11 | 1466 | 84 | |
| JP042596.1 | - | 10 - 11 | 1466 | 84 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map