Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 93 - 94 | 21606 | 65 | |
| XM_007645556.1 | XP_007643746.1 | 93 - 94 | 21606 | 65 | ||
| XM_007645557.1 | XP_007643747.1 | 92 - 93 | 21543 | 65 | ||
| XM_007645558.1 | XP_007643748.1 | 88 - 89 | 15510 | 65 | ||
| XM_007645559.1 | XP_007643749.1 | 88 - 89 | 15510 | 65 | ||
| XM_007645560.1 | XP_007643750.1 | 87 - 88 | 15447 | 65 | ||
| XM_007645561.1 | XP_007643751.1 | 37 - 38 | 10234 | 65 | ||
| GenBank mRNA | JI890392.1 | - | 11 - 12 | 1621 | 65 | |
| JP042596.1 | - | 11 - 12 | 1621 | 65 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map