Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 96 - 97 | 21816 | 110 | |
| XM_007645556.1 | XP_007643746.1 | 95 - 96 | 21798 | 110 | ||
| XM_007645557.1 | XP_007643747.1 | 95 - 96 | 21753 | 110 | ||
| XM_007645558.1 | XP_007643748.1 | 91 - 92 | 15720 | 110 | ||
| XM_007645559.1 | XP_007643749.1 | 90 - 91 | 15702 | 110 | ||
| XM_007645560.1 | XP_007643750.1 | 89 - 90 | 15639 | 110 | ||
| XM_007645561.1 | XP_007643751.1 | 40 - 41 | 10444 | 110 | ||
| GenBank mRNA | JI890392.1 | - | 13 - 14 | 1813 | 110 | |
| JP042596.1 | - | 13 - 14 | 1813 | 110 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map