Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 97 - 98 | 22028 | 77 | |
| XM_007645556.1 | XP_007643746.1 | 96 - 97 | 22010 | 77 | ||
| XM_007645557.1 | XP_007643747.1 | 96 - 97 | 21965 | 77 | ||
| XM_007645558.1 | XP_007643748.1 | 92 - 93 | 15932 | 77 | ||
| XM_007645559.1 | XP_007643749.1 | 91 - 92 | 15914 | 77 | ||
| XM_007645560.1 | XP_007643750.1 | 90 - 91 | 15851 | 77 | ||
| XM_007645561.1 | XP_007643751.1 | 41 - 42 | 10656 | 77 | ||
| GenBank mRNA | JI890392.1 | - | 14 - 15 | 2025 | 77 | |
| JP042596.1 | - | 14 - 15 | 2025 | 77 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map