Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 61 - 62 | 16392 | 83 | |
| XM_007645556.1 | XP_007643746.1 | 61 - 62 | 16392 | 83 | ||
| XM_007645557.1 | XP_007643747.1 | 60 - 61 | 16329 | 83 | ||
| XM_007645558.1 | XP_007643748.1 | 56 - 57 | 10296 | 83 | ||
| XM_007645559.1 | XP_007643749.1 | 56 - 57 | 10296 | 83 | ||
| XM_007645560.1 | XP_007643750.1 | 55 - 56 | 10233 | 83 | ||
| XM_007645561.1 | XP_007643751.1 | 5 - 6 | 5020 | 83 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map