Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 63 - 64 | 16806 | 89 | |
| XM_007645556.1 | XP_007643746.1 | 63 - 64 | 16806 | 89 | ||
| XM_007645557.1 | XP_007643747.1 | 62 - 63 | 16743 | 89 | ||
| XM_007645558.1 | XP_007643748.1 | 58 - 59 | 10710 | 89 | ||
| XM_007645559.1 | XP_007643749.1 | 58 - 59 | 10710 | 89 | ||
| XM_007645560.1 | XP_007643750.1 | 57 - 58 | 10647 | 89 | ||
| XM_007645561.1 | XP_007643751.1 | 7 - 8 | 5434 | 89 | ||
| GenBank mRNA | JI895671.1 | - | 2 - 3 | 191 | 89 | |
| JP043055.1 | - | 2 - 3 | 191 | 89 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map