Assay Details
| Gene Symbol: LOC100770726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645555.1 | XP_007643745.1 | 64 - 65 | 17053 | 64 | |
| XM_007645556.1 | XP_007643746.1 | 64 - 65 | 17053 | 64 | ||
| XM_007645557.1 | XP_007643747.1 | 63 - 64 | 16990 | 64 | ||
| XM_007645558.1 | XP_007643748.1 | 59 - 60 | 10957 | 64 | ||
| XM_007645559.1 | XP_007643749.1 | 59 - 60 | 10957 | 64 | ||
| XM_007645560.1 | XP_007643750.1 | 58 - 59 | 10894 | 64 | ||
| XM_007645561.1 | XP_007643751.1 | 8 - 9 | 5681 | 64 | ||
| GenBank mRNA | JI895671.1 | - | 3 - 4 | 438 | 64 | |
| JP043055.1 | - | 3 - 4 | 438 | 64 | ||
More Information
Set Membership: |
|

LOC100770726
Genomic Map