Assay Details
Gene Symbol: Pmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510075.1 | XP_003510123.1 | 2 - 3 | 245 | 62 | |
XM_007650718.1 | XP_007648908.1 | 2 - 3 | 245 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
