Assay Details
| Gene Symbol: Rpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499579.2 | XP_003499627.1 | 3 - 4 | 362 | 60 | |
| GenBank mRNA | JI867826.1 | - | 3 - 4 | 267 | 60 | |
More Information
Set Membership: |
|

Rpl19
Genomic Map