Assay Details
| Gene Symbol: Pcyox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507976.2 | XP_003508024.1 | 2 - 3 | 362 | 60 | |
| XM_007648563.1 | XP_007646753.1 | 2 - 3 | 479 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007648563.1 transcripts.
Set Membership: |
|

Pcyox1
Genomic Map