Assay Details
Gene Symbol: Gabpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007631367.1 | XP_007629557.1 | 7 - 8 | 1171 | 62 | |
XM_007631372.1 | XP_007629562.1 | 8 - 9 | 1234 | 62 | ||
XM_007631381.1 | XP_007629571.1 | 7 - 8 | 1500 | 62 | ||
XM_007631385.1 | XP_007629575.1 | 7 - 8 | 1471 | 62 | ||
XM_007631391.1 | XP_007629581.1 | 7 - 8 | 983 | 62 | ||
GenBank mRNA | JI886160.1 | - | 2 - 3 | 312 | 62 | |
JP038254.1 | - | 2 - 3 | 312 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
