Assay Details
| Gene Symbol: Gpr126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641869.1 | XP_007640059.1 | 23 - 24 | 3637 | 80 | |
| XM_007641870.1 | XP_007640060.1 | 22 - 23 | 3553 | 80 | ||
More Information
Set Membership: |
|

Gpr126
Genomic Map