Assay Details
| Gene Symbol: Dfna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498555.2 | XP_003498603.1 | 3 - 4 | 830 | 57 | |
| XM_007629677.1 | XP_007627867.1 | 3 - 4 | 792 | 57 | ||
| XM_007629684.1 | XP_007627874.1 | 4 - 5 | 1569 | 57 | ||
| XM_007629697.1 | XP_007627887.1 | 3 - 4 | 497 | 57 | ||
| XM_007629703.1 | XP_007627893.1 | 3 - 4 | 448 | 57 | ||
| XM_007629710.1 | XP_007627900.1 | 3 - 4 | 1260 | 57 | ||
More Information
Set Membership: |
|

Dfna5
Genomic Map