Assay Details
| Gene Symbol: Ribc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500197.2 | XP_003500245.2 | 4 - 5 | 591 | 57 | |
| XM_007640945.1 | XP_007639135.1 | 4 - 5 | 700 | 57 | ||
| XM_007640946.1 | XP_007639136.1 | 4 - 5 | 661 | 57 | ||
| XM_007640947.1 | XP_007639137.1 | 4 - 5 | 622 | 57 | ||
| GenBank mRNA | JI880463.1 | - | 1 - 2 | 260 | 57 | |
More Information
Set Membership: |
|

Ribc1
Genomic Map