Assay Details
| Gene Symbol: LOC103164583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007655173.1 | XP_007653363.1 | 1 - 2 | 30 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103164583| Gene Symbol: LOC103164583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007655173.1 | XP_007653363.1 | 1 - 2 | 30 | 58 | |
Set Membership: |
|