Assay Details
| Gene Symbol: Abhd17c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511699.1 | XP_003511747.1 | 1 - 2 | 228 | 106 | |
| GenBank mRNA | JI872939.1 | - | 1 - 2 | 243 | 106 | |
| JP056843.1 | - | 1 - 2 | 243 | 106 | ||
More Information
Set Membership: |
|

Abhd17c
Genomic Map