Assay Details
| Gene Symbol: LOC103158748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607848.1 | XP_007606038.1 | 5 - 6 | 860 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103158748| Gene Symbol: LOC103158748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607848.1 | XP_007606038.1 | 5 - 6 | 860 | 71 | |
Set Membership: |
|