Assay Details
| Gene Symbol: Lrrc47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654291.1 | XP_007652481.1 | 2 - 3 | 500 | 68 | |
| GenBank mRNA | JI866814.1 | - | 1 - 2 | 189 | 68 | |
| JP059676.1 | - | 1 - 2 | 189 | 68 | ||
More Information
Set Membership: |
|

Lrrc47
Genomic Map