Assay Details
| Gene Symbol: Lcorl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646466.1 | XP_007644656.1 | 4 - 5 | 518 | 89 | |
| XM_007646467.1 | XP_007644657.1 | 4 - 5 | 518 | 89 | ||
| XM_007646468.1 | XP_007644658.1 | 4 - 5 | 518 | 89 | ||
| XM_007646469.1 | XP_007644659.1 | 4 - 5 | 518 | 89 | ||
| XM_007646470.1 | XP_007644660.1 | 3 - 4 | 440 | 89 | ||
| XM_007646471.1 | XP_007644661.1 | 4 - 5 | 520 | 89 | ||
| XM_007646472.1 | XP_007644662.1 | 4 - 5 | 520 | 89 | ||
More Information
Set Membership: |
|

Lcorl
Genomic Map