Assay Details
| Gene Symbol: Rest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511484.2 | XP_003511532.2 | 2 - 3 | 1023 | 93 | |
| XM_007652020.1 | XP_007650210.1 | 2 - 3 | 1023 | 93 | ||
| XM_007652021.1 | XP_007650211.1 | 2 - 3 | 1023 | 93 | ||
More Information
Set Membership: |
|

Rest
Genomic Map