Assay Details
| Gene Symbol: Lrrc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498045.2 | XP_003498093.1 | 4 - 5 | 617 | 75 | |
| XM_007621941.1 | XP_007620131.1 | 4 - 5 | 757 | 75 | ||
| XM_007621948.1 | XP_007620138.1 | 4 - 5 | 564 | 75 | ||
| GenBank mRNA | JI873112.1 | - | 1 - 2 | 75 | ||
More Information
Set Membership: |
|

Lrrc57
Genomic Map