Assay Details
| Gene Symbol: Wbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510925.2 | XP_003510973.1 | 3 - 4 | 400 | 92 | |
| XM_007651509.1 | XP_007649699.1 | 4 - 5 | 509 | 92 | ||
| XM_007651510.1 | XP_007649700.1 | 4 - 5 | 553 | 92 | ||
More Information
Set Membership: |
|

Wbp1l
Genomic Map