Assay Details
| Gene Symbol: Hyi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643286.1 | XP_007641476.1 | 4 - 5 | 363 | 59 | |
| GenBank mRNA | JI862659.1 | - | 4 - 5 | 242 | 59 | |
More Information
Set Membership: |
|

Hyi
Genomic Map