Assay Details
| Gene Symbol: Tlcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513830.2 | XP_003513878.1 | 3 - 4 | 442 | 86 | |
| XM_007654032.1 | XP_007652222.1 | 3 - 4 | 620 | 86 | ||
| XM_007654033.1 | XP_007652223.1 | 3 - 4 | 238 | 86 | ||
| GenBank mRNA | JI891898.1 | - | 3 - 4 | 276 | 86 | |
| JP041838.1 | - | 3 - 4 | 275 | 86 | ||
More Information
Set Membership: |
|

Tlcd1
Genomic Map