Assay Details
| Gene Symbol: LOC100773076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644322.1 | XP_007642512.1 | 1 - 2 | 273 | 77 | |
| GenBank mRNA | JI884369.1 | - | 2 - 3 | 510 | 77 | |
More Information
Set Membership: |
|

LOC100773076
Genomic Map