Assay Details
| Gene Symbol: Itm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507558.1 | XP_003507606.1 | 3 - 4 | 436 | 65 | |
| XM_003507559.1 | XP_003507607.1 | 2 - 3 | 304 | 65 | ||
More Information
Set Membership: |
|

Itm2a
Genomic Map