Assay Details
| Gene Symbol: Ereg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512370.2 | XP_003512418.1 | 3 - 4 | 459 | 69 | |
| GenBank mRNA | JI887722.1 | - | 3 - 4 | 397 | 69 | |
| JP058604.1 | - | 3 - 4 | 397 | 69 | ||
More Information
Set Membership: |
|

Ereg
Genomic Map