Assay Details
| Gene Symbol: Ppp1r1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502134.1 | XP_003502182.1 | 3 - 4 | 185 | 67 | |
| XM_007642765.1 | XP_007640955.1 | 3 - 4 | 206 | 67 | ||
More Information
Set Membership: |
|

Ppp1r1c
Genomic Map