Assay Details
| Gene Symbol: Sod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507569.2 | XP_003507617.1 | 4 - 5 | 613 | 54 | |
| GenBank mRNA | JI889465.1 | - | 3 - 4 | 506 | 54 | |
More Information
Set Membership: |
|

Sod2
Genomic Map