Assay Details
| Gene Symbol: Bend4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513778.2 | XP_003513826.1 | 3 - 4 | 932 | 82 | |
| GenBank mRNA | JI886972.1 | - | 3 - 4 | 868 | 82 | |
| JP059225.1 | - | 3 - 4 | 868 | 82 | ||
More Information
Set Membership: |
|

Bend4
Genomic Map