Assay Details
Gene Symbol: Il25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646594.1 | XP_007644784.1 | 3 - 4 | 1037 | 64 | |
XM_007646595.1 | XP_007644785.1 | 3 - 4 | 1126 | 64 | ||
XM_007646596.1 | XP_007644786.1 | 3 - 4 | 421 | 64 | ||
XM_007646597.1 | XP_007644787.1 | 3 - 4 | 1101 | 64 |

More Information
Set Membership: |
![]() ![]() |
