Assay Details
| Gene Symbol: Atg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503726.2 | XP_003503774.1 | 7 - 8 | 801 | 66 | |
| XM_007644298.1 | XP_007642488.1 | 8 - 9 | 880 | 66 | ||
| XM_007644299.1 | XP_007642489.1 | 8 - 9 | 771 | 66 | ||
| XM_007644300.1 | XP_007642490.1 | 8 - 9 | 769 | 66 | ||
| GenBank mRNA | JI880639.1 | - | 3 - 4 | 316 | 66 | |
More Information
Set Membership: |
|

Atg10
Genomic Map