Assay Details
| Gene Symbol: Sage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640578.1 | XP_007638768.1 | 3 - 4 | 427 | 141 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sage1| Gene Symbol: Sage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640578.1 | XP_007638768.1 | 3 - 4 | 427 | 141 | |
Set Membership: |
|