Assay Details
| Gene Symbol: Il4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651465.1 | XP_007649655.1 | 6 - 7 | 827 | 70 | |
| XM_007651466.1 | XP_007649656.1 | 6 - 7 | 827 | 70 | ||
| XM_007651467.1 | XP_007649657.1 | 4 - 5 | 512 | 70 | ||
More Information
Set Membership: |
|

Il4i1
Genomic Map