Assay Details
| Gene Symbol: LOC100773524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507417.2 | XP_003507465.1 | 2 - 3 | 713 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100773524| Gene Symbol: LOC100773524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507417.2 | XP_003507465.1 | 2 - 3 | 713 | 75 | |
Set Membership: |
|