Assay Details
| Gene Symbol: LOC100764703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512049.2 | XP_003512097.1 | 2 - 3 | 482 | 67 | |
| GenBank mRNA | JP047303.1 | - | 2 - 3 | 423 | 67 | |
More Information
Set Membership: |
|

LOC100764703
Genomic Map